COMH4211A4A8 Combi-Blocks, Inc.

3-Methyl-4-(propane-1-sulfonyl)-benzoic acid

  • MDL number MFCD31579486
  • Cas Number : 1857293-46-9
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0069

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0069
Availability details for
Quantity
  • Availability
  • Add to Cart