COMH4211A4AA Combi-Blocks, Inc.

N-Boc-o-allyl-n-benzyl hydroxylamine

  • MDL number MFCD31628734
  • Cas Number : 2270909-51-6
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0081

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0081
Availability details for
Quantity
  • Availability
  • Add to Cart