COMH4211A4AF Combi-Blocks, Inc.

3-(1-Phenyl-ethylsulfanyl)-propionic acid

  • MDL number MFCD19447266
  • Cas Number : 93664-13-2
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0090

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0090
Availability details for
Quantity
  • Availability
  • Add to Cart