COMH4211A4B0 Combi-Blocks, Inc.

(5-Bromo-2-methyl-phenyl)-pyrrolidin-1-yl-methanone

  • MDL number MFCD21508340
  • Cas Number : 1480283-73-5
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0091

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0091
Availability details for
Quantity
  • Availability
  • Add to Cart