COMH4211A4B1 Combi-Blocks, Inc.

Ethyl 3-(3-amino-2-methylphenyl)propanoate

  • MDL number MFCD24627125
  • Cas Number : 757179-63-8
  • Molecular Weight : 207.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0093

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0093
Availability details for
Quantity
  • Availability
  • Add to Cart