COMH4211A4B2 Combi-Blocks, Inc.

3-Cyclobutylphenol

  • MDL number MFCD21099817
  • Cas Number : 211495-49-7
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-cyclobutylphenol
Inchi Code 1S/C10H12O/c11-10-6-2-5-9(7-10)8-3-1-4-8/h2,5-8,11H,1,3-4H2
Inchi Key KGAOBXXXNFRSSW-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0096

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0096
Availability details for
Quantity
  • Availability
  • Add to Cart