COMH4211A4B3 Combi-Blocks, Inc.

1-[2-Chloro-4-(trifluoromethyl)phenyl]ethan-1-amine

  • MDL number MFCD24368262
  • Cas Number : 1337041-66-3
  • Molecular Weight : 223.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0100

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0100
Availability details for
Quantity
  • Availability
  • Add to Cart