COMH4211A4B4 Combi-Blocks, Inc.

4-Bromo-3-ethoxy-n,n-diethylbenzamide

  • MDL number MFCD30830147
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0101

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0101
Availability details for
Quantity
  • Availability
  • Add to Cart