COMH4211A4B5 Combi-Blocks, Inc.

Ethyl 2-formylcinnamate

  • MDL number MFCD03789578
  • Cas Number : 86867-62-1
  • Molecular Weight : 204.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0102
Availability details for
Quantity
  • Availability
  • Add to Cart