COMH4211A4B6 Combi-Blocks, Inc.

2-(Chroman-4-yloxy)acetic acid

  • MDL number MFCD24502473
  • Cas Number : 1359734-98-7
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0106
Availability details for
Quantity
  • Availability
  • Add to Cart