COMH4211A4B7 Combi-Blocks, Inc.

[1-(2-Bromo-5-trifluoromethyl-phenyl)-ethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD31539538
  • Cas Number : 2204588-08-7
  • Molecular Weight : 368.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0107

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0107
Availability details for
Quantity
  • Availability
  • Add to Cart