COMH4211A4B8 Combi-Blocks, Inc.

N,N-Bis(4-methoxybenzyl)methanesulfonamide

  • MDL number MFCD31613785
  • Cas Number : 852358-71-5
  • Molecular Weight : 335.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0115

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0115
Availability details for
Quantity
  • Availability
  • Add to Cart