COMH4211A4B9 Combi-Blocks, Inc.

4,4-Dimethoxy-1-(1,3-thiazol-2-yl)butane-1,3-dione

  • MDL number MFCD21143813
  • Cas Number : 1482564-21-5
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0122

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0122
Availability details for
Quantity
  • Availability
  • Add to Cart