COMH4211A4BA Combi-Blocks, Inc.

5-(2-Bromo-5-nitrophenyl)oxazole

  • MDL number MFCD31614014
  • Cas Number : 1638604-66-6
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0127

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0127
Availability details for
Quantity
  • Availability
  • Add to Cart