COMH4211A4BB Combi-Blocks, Inc.

2-(Ethylsulfonyl)-5-nitrobenzonitrile

  • MDL number MFCD27932796
  • Cas Number : 918810-27-2
  • Molecular Weight : 240.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(ethylsulfonyl)-5-nitrobenzonitrile
Inchi Code 1S/C9H8N2O4S/c1-2-16(14,15)9-4-3-8(11(12)13)5-7(9)6-10/h3-5H,2H2,1H3
Inchi Key GOMGTLCRLJAJFE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0128

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0128
Availability details for
Quantity
  • Availability
  • Add to Cart