COMH4211A4BC Combi-Blocks, Inc.

3-(2-Fluoro-4-methyl-phenyl)-2-methyl-propan-1-ol

  • MDL number MFCD31579329
  • Cas Number : 2145856-41-1
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0129

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0129
Availability details for
Quantity
  • Availability
  • Add to Cart