COMH4211A4BD Combi-Blocks, Inc.

4-Hydroxy-pyrazole-1-carboxylic acid methylamide

  • MDL number MFCD31583101
  • Cas Number : 2270905-75-2
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydroxy-N-methyl-1H-pyrazole-1-carboxamide
Inchi Code 1S/C5H7N3O2/c1-6-5(10)8-3-4(9)2-7-8/h2-3,9H,1H3,(H,6,10)
Inchi Key GCWGGXANWDCGLG-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0131

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0131
Availability details for
Quantity
  • Availability
  • Add to Cart