COMH4211A4BE Combi-Blocks, Inc.

(1R,3R)-3-(((Benzyloxy)carbonyl)amino)cyclohexanecarboxylic acid

  • MDL number MFCD28344088
  • Cas Number : 1035325-24-6
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0132

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0132
Availability details for
Quantity
  • Availability
  • Add to Cart