COMH4211A4BF Combi-Blocks, Inc.

2-(2-Cyclopropylmethoxy-phenyl)-ethanol

  • MDL number MFCD31628782
  • Cas Number : 1384452-88-3
  • Molecular Weight : 192.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0133

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0133
Availability details for
Quantity
  • Availability
  • Add to Cart