COMH4211A4C1 Combi-Blocks, Inc.

2-[(3,5-Dimethylphenyl)formamido]propanoic acid

  • MDL number MFCD09836861
  • Cas Number : 1423044-65-8
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0135

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0135
Availability details for
Quantity
  • Availability
  • Add to Cart