COMH4211A4C2 Combi-Blocks, Inc.

5-(4-Ethoxy-2-fluoro-phenyl)-1h-tetrazole

  • MDL number MFCD31579403
  • Cas Number : 2096447-81-1
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0136

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0136
Availability details for
Quantity
  • Availability
  • Add to Cart