COMH4211A4C3 Combi-Blocks, Inc.

1,2-O-Isopropylidene-beta-l-arabinoftranose

  • MDL number MFCD31583345
  • Cas Number : 51885-64-4
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0141

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0141
Availability details for
Quantity
  • Availability
  • Add to Cart