COMH4211A4C4 Combi-Blocks, Inc.

8,8-Bis((2-carboxyethoxy)methyl)-3,6-dioxo-1-phenyl-2,10-dioxa-4,7-diazatridecan-13-oic acid

  • MDL number MFCD18252296
  • Cas Number : 220886-02-2
  • Molecular Weight : 528.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0142

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0142
Availability details for
Quantity
  • Availability
  • Add to Cart