COMH4211A4C7 Combi-Blocks, Inc.

2-Ethynyl-4-fluoro-1-methylbenzene

  • MDL number MFCD11618092
  • Cas Number : 1233512-88-3
  • Molecular Weight : 134.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0148

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0148
Availability details for
Quantity
  • Availability
  • Add to Cart