COMH4211A4C9 Combi-Blocks, Inc.

4-(4-Hydroxymethyl-phenoxy)-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD28965642
  • Cas Number : 1203600-49-0
  • Molecular Weight : 341.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 4-(4-(hydroxymethyl)phenoxy)piperidine-1-carboxylate
Inchi Code 1S/C20H23NO4/c22-14-16-6-8-18(9-7-16)25-19-10-12-21(13-11-19)20(23)24-15-17-4-2-1-3-5-17/h1-9,19,22H,10-15H2
Inchi Key DPJPNPSCNJUTEA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0152

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0152
Availability details for
Quantity
  • Availability
  • Add to Cart