COMH4211A4CA Combi-Blocks, Inc.

5-Bromo-n,n,2-trimethylbenzamide

  • MDL number MFCD21193021
  • Cas Number : 908257-16-9
  • Molecular Weight : 242.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0154

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0154
Availability details for
Quantity
  • Availability
  • Add to Cart