COMH4211A4CB Combi-Blocks, Inc.

Dehydrodeoxy donepezil

  • MDL number MFCD16294189
  • Cas Number : 120013-45-8
  • Molecular Weight : 363.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0155

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0155
Availability details for
Quantity
  • Availability
  • Add to Cart