COMH4211A4CE Combi-Blocks, Inc.

Trans-4-hydroxycyclohexane-1-carboxylic acid benzyl ester

  • MDL number MFCD28383626
  • Cas Number : 933448-77-2
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0159

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0159
Availability details for
Quantity
  • Availability
  • Add to Cart