COMH4211A4D2 Combi-Blocks, Inc.

3-(3-Cyclopropylmethoxy-phenyl)-propionic acid

  • MDL number MFCD31561585
  • Cas Number : 1350515-10-4
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0166

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0166
Availability details for
Quantity
  • Availability
  • Add to Cart