COMH4211A4D3 Combi-Blocks, Inc.

Methyl 2-(4-(4-hydroxyphenyl)cyclohexyl)acetate

  • MDL number MFCD31561586
  • Cas Number : 1245708-05-7
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0172

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0172
Availability details for
Quantity
  • Availability
  • Add to Cart