COMH4211A4D7 Combi-Blocks, Inc.

C-(4-Isopropoxy-phenyl)-c-phenyl-methylamine hydrochloride

  • MDL number MFCD31561549
  • Cas Number : 2270906-28-8
  • Molecular Weight : 277.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-isopropoxyphenyl)(phenyl)methanamine hydrochloride
Inchi Code 1S/C16H19NO.ClH/c1-12(2)18-15-10-8-14(9-11-15)16(17)13-6-4-3-5-7-13;/h3-12,16H,17H2,1-2H3;1H
Inchi Key HIIPWTTYYNMWFD-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0177

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0177
Availability details for
Quantity
  • Availability
  • Add to Cart