COMH4211A4D8 Combi-Blocks, Inc.

3-Bromo-4-(4-methoxy-benzyloxy)-phenylamine

  • MDL number MFCD16512147
  • Cas Number : 1290716-13-0
  • Molecular Weight : 308.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0179

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0179
Availability details for
Quantity
  • Availability
  • Add to Cart