COMH4211A4D9 Combi-Blocks, Inc.

Carbonic acid cyclopropylmethyl ester 4-nitro-phenyl ester

  • MDL number MFCD31583125
  • Cas Number : 276697-73-5
  • Molecular Weight : 237.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0182

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0182
Availability details for
Quantity
  • Availability
  • Add to Cart