COMH4211A4DB Combi-Blocks, Inc.

4-(4-Amino-phenylsulfanylmethyl)-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD31614093
  • Cas Number : 2270907-15-6
  • Molecular Weight : 356.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 4-(((4-aminophenyl)thio)methyl)piperidine-1-carboxylate
Inchi Code 1S/C20H24N2O2S/c21-18-6-8-19(9-7-18)25-15-17-10-12-22(13-11-17)20(23)24-14-16-4-2-1-3-5-16/h1-9,17H,10-15,21H2
Inchi Key CMKIFUYJPXFZIN-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0187

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0187
Availability details for
Quantity
  • Availability
  • Add to Cart