COMH4211A4DD Combi-Blocks, Inc.

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(1-(3-(trifluoro-methyl)phenyl)ethyl)-1h-pyrazole

  • MDL number MFCD31583088
  • Cas Number : 1604036-71-6
  • Molecular Weight : 366.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0191

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0191
Availability details for
Quantity
  • Availability
  • Add to Cart