COMH4211A4E0 Combi-Blocks, Inc.

4-(4-Methoxybenzyloxy)-3-chlorobenzaldehyde

  • MDL number MFCD13807436
  • Cas Number : 851402-45-4
  • Molecular Weight : 276.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-4-[(4-methoxybenzyl)oxy]benzaldehyde
Inchi Code 1S/C15H13ClO3/c1-18-13-5-2-11(3-6-13)10-19-15-7-4-12(9-17)8-14(15)16/h2-9H,10H2,1H3
Inchi Key ZCSMXEZZMBGPBA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0196

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0196
Availability details for
Quantity
  • Availability
  • Add to Cart