COMH4211A4FC Combi-Blocks, Inc.

N-(2,4-Dimethoxybenzyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD19428269
  • Cas Number : 1235406-58-2
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0199

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0199
Availability details for
Quantity
  • Availability
  • Add to Cart