COMH4211A4FD Combi-Blocks, Inc.

2-[4-(2-Chloro-phenyl)-pyrazol-1-yl]-ethanol

  • MDL number MFCD19319109
  • Cas Number : 1267158-10-0
  • Molecular Weight : 222.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0201

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0201
Availability details for
Quantity
  • Availability
  • Add to Cart