COMH4211A4FF Combi-Blocks, Inc.

2-Chloro-4-isopropoxy-5-nitro-benzoic acid

  • MDL number MFCD28974505
  • Cas Number : 1032903-53-9
  • Molecular Weight : 259.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0208

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0208
Availability details for
Quantity
  • Availability
  • Add to Cart