COMH4211A502 Combi-Blocks, Inc.

4-(4-Fluoro-3-methoxyphenyl)butanoic acid

  • MDL number MFCD31561457
  • Cas Number : 1517035-68-5
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0217

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0217
Availability details for
Quantity
  • Availability
  • Add to Cart