COMH4211A503 Combi-Blocks, Inc.

4-(3-Fluoro-4-methoxycarbonyl-henyl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31583075
  • Cas Number : 1488388-73-3
  • Molecular Weight : 337.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0221

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0221
Availability details for
Quantity
  • Availability
  • Add to Cart