COMH4211A937 Combi-Blocks, Inc.

4,4-Difluoro-1-(3-trifluoromethyl-phenyl)-butane-1,3-dione

  • MDL number MFCD11541445
  • Cas Number : 911112-56-6
  • Molecular Weight : 266.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0222

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0222
Availability details for
Quantity
  • Availability
  • Add to Cart