COMH4211A938 Combi-Blocks, Inc.

(2S)-Glycidyl 1-methylindol-4-yl ether

  • MDL number MFCD31614111
  • Cas Number : 256372-09-5
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0224
Availability details for
Quantity
  • Availability
  • Add to Cart