COMH4211A939 Combi-Blocks, Inc.

1-(3-Benzyloxy-phenyl)-cyclobutanol

  • MDL number MFCD31613897
  • Cas Number : 211495-48-6
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0226

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0226
Availability details for
Quantity
  • Availability
  • Add to Cart