COMH4211A93A Combi-Blocks, Inc.

(5-Bromo-2-chloro-pyridin-3-yl)-(4-ethyl-piperazin-1-yl)-methanone

  • MDL number MFCD15504501
  • Cas Number : 1458679-34-9
  • Molecular Weight : 332.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0227

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0227
Availability details for
Quantity
  • Availability
  • Add to Cart