COMH4211A93D Combi-Blocks, Inc.

N-(2-(4-(Trifluoromethoxy)phenoxy)phenyl)piperidine-4-carboxamide

  • MDL number MFCD30557300
  • Cas Number : 919117-95-6
  • Molecular Weight : 380.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0232

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0232
Availability details for
Quantity
  • Availability
  • Add to Cart