COMH4211A93F Combi-Blocks, Inc.

Tri-tert-butyl 2,2',2''-(10-(2-((2,5-dioxopyrrolidin-1-yl)oxy)-2-oxoethyl)-1,4,7,10-tetraazacyclododecane-1,4,7-triyl)triacetate

  • MDL number MFCD26131244
  • Cas Number : 819869-77-7
  • Molecular Weight : 669.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0236

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0236
Availability details for
Quantity
  • Availability
  • Add to Cart