COMH4211A941 Combi-Blocks, Inc.

4-(6-Bromo-7-methylimidazo[1,2-a]pyridin-3-yl)benzonitrile

  • MDL number MFCD31583204
  • Cas Number : 1464156-15-7
  • Molecular Weight : 312.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0239

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0239
Availability details for
Quantity
  • Availability
  • Add to Cart