COMH4211A944 Combi-Blocks, Inc.

3-(4-Fluorophenyl)-4-isoxazolecarboxylic acid ethyl ester

  • MDL number MFCD00981276
  • Cas Number : 159693-06-8
  • Molecular Weight : 235.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0247

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0247
Availability details for
Quantity
  • Availability
  • Add to Cart