COMH4211A945 Combi-Blocks, Inc.

4-(3-Fluoro-4-nitro-phenoxy)-piperidine hydrochloride

  • MDL number MFCD31613960
  • Cas Number : 1660980-17-5
  • Molecular Weight : 276.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0250

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0250
Availability details for
Quantity
  • Availability
  • Add to Cart